siRNA Details
| Gene Symbol: Prmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024363.1 | NP_077339.1 | 9, 10 | 985 | ||
| XM_006229144.2 | XP_006229206.1 | 10, 11 | 1103 | |||
| XM_006229145.2 | XP_006229207.1 | 9, 10 | 1267 | |||
| XM_006229146.2 | XP_006229208.1 | 10, 11 | 1370 | |||
| GenBank mRNA | BC078815.1 | - | 9, 10 | 990 | 7106257 | |
| FQ213198.1 | - | 9, 10 | 998 | |||
| FQ219843.1 | - | 9, 10 | 996 | |||
| FQ220982.1 | - | 7, 8 | 737 | |||
| FQ228625.1 | - | 9, 10 | 1266 | |||
| FQ233897.1 | - | 9, 10 | 1000 | |||
| U60882.1 | - | 9, 10 | 985 | |||

Prmt1
Genomic Map