siRNA Details
| Gene Symbol: Slc28a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031664.1 | NP_113852.1 | 4 | 478 | ||
| GenBank mRNA | AY029302.1 | - | 4 | 459 | ||
| EU032627.2 | - | 4 | 546 | |||
| U25055.1 | - | 4 | 478 | |||
| U66723.1 | - | 4 | 449 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc28a2| Gene Symbol: Slc28a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031664.1 | NP_113852.1 | 4 | 478 | ||
| GenBank mRNA | AY029302.1 | - | 4 | 459 | ||
| EU032627.2 | - | 4 | 546 | |||
| U25055.1 | - | 4 | 478 | |||
| U66723.1 | - | 4 | 449 | |||