siRNA Details
| Gene Symbol: Kitlg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021843.4 | NP_068615.1 | 6, 7 | 772 | ||
| XM_008765339.1 | XP_008763561.1 | 6, 7 | 816 | |||
| GenBank mRNA | AF071204.1 | - | 6, 7 | 607 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013598, S40536, X99322, M64262, U44725, and others.

Kitlg
Genomic Map