siRNA Details
| Gene Symbol: Rfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021849.1 | NP_068621.1 | 7 | 901 | ||
| XM_006247910.2 | XP_006247972.1 | 7 | 967 | |||
| XM_008768480.1 | XP_008766702.1 | 7 | 1024 | |||
| XM_008768481.1 | XP_008766703.1 | 7 | 1024 | |||
| GenBank mRNA | AB016486.1 | - | 7 | 901 | ||
| BC062031.1 | - | 7 | 946 | 5623611 | ||

Rfng
Genomic Map