siRNA Details
| Gene Symbol: Clock | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289832.1 | NP_001276761.1 | 18 | 2099 | ||
| NM_021856.2 | NP_068628.1 | 19 | 2189 | |||
| XM_006250869.2 | XP_006250931.1 | 19 | 2236 | |||
| XM_006250870.2 | XP_006250932.1 | 19 | 2233 | |||
| GenBank mRNA | AB019258.1 | - | 19 | 2189 | ||
| AB019259.1 | - | 18 | 2099 | |||

Clock
Genomic Map