siRNA Details
| Gene Symbol: Gnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021858.3 | NP_068630.1 | 8, 9 | 806 | ||
| GenBank mRNA | BC086422.1 | - | 8, 9 | 809 | 7193131 | |
| DQ120487.1 | - | 6, 7 | 616 | |||
| DQ120488.1 | - | 6, 7 | 616 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnb3| Gene Symbol: Gnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021858.3 | NP_068630.1 | 8, 9 | 806 | ||
| GenBank mRNA | BC086422.1 | - | 8, 9 | 809 | 7193131 | |
| DQ120487.1 | - | 6, 7 | 616 | |||
| DQ120488.1 | - | 6, 7 | 616 | |||