siRNA Details
Gene Symbol: Batf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021865.1 | NP_068637.1 | 3 | 302 | ||
XM_008769873.1 | XP_008768095.1 | 3 | 414 | |||
XM_008769874.1 | XP_008768096.1 | 3 | 442 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Batf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021865.1 | NP_068637.1 | 3 | 302 | ||
XM_008769873.1 | XP_008768095.1 | 3 | 414 | |||
XM_008769874.1 | XP_008768096.1 | 3 | 442 |