siRNA Details
| Gene Symbol: Syt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022191.1 | NP_071527.1 | 5 | 1566 | ||
| GenBank mRNA | BC099185.1 | - | 1222 | 7383303 | ||
| U20105.1 | - | 5 | 1566 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syt6| Gene Symbol: Syt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022191.1 | NP_071527.1 | 5 | 1566 | ||
| GenBank mRNA | BC099185.1 | - | 1222 | 7383303 | ||
| U20105.1 | - | 5 | 1566 | |||