siRNA Details
| Gene Symbol: Treh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136141.1 | NP_001129613.1 | 12 | 1350 | ||
| XM_006242956.2 | XP_006243018.1 | 11 | 1222 | |||
| GenBank mRNA | AF038043.1 | - | 11 | 1271 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Treh| Gene Symbol: Treh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136141.1 | NP_001129613.1 | 12 | 1350 | ||
| XM_006242956.2 | XP_006243018.1 | 11 | 1222 | |||
| GenBank mRNA | AF038043.1 | - | 11 | 1271 | ||