siRNA Details
| Gene Symbol: Slc15a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031672.2 | NP_113860.2 | 11 | 1145 | ||
| GenBank mRNA | BC085327.1 | - | 11 | 1145 | 7098404 | |
| D63149.1 | - | 11 | 1133 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc15a2| Gene Symbol: Slc15a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031672.2 | NP_113860.2 | 11 | 1145 | ||
| GenBank mRNA | BC085327.1 | - | 11 | 1145 | 7098404 | |
| D63149.1 | - | 11 | 1133 | |||