siRNA Details
| Gene Symbol: Il1rn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022194.2 | NP_071530.1 | 2 | 148 | ||
| XM_006233636.2 | XP_006233698.1 | 3 | 2391 | |||
| XM_006233637.2 | XP_006233699.1 | 3 | 178 | |||
| XM_006233638.2 | XP_006233700.1 | 2 | 78 | |||
| GenBank mRNA | BC070930.1 | - | 2 | 142 | ||
| CK364274.1 | - | 2 | 172 | |||

Il1rn
Genomic Map