siRNA Details
| Gene Symbol: Ivl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022195.1 | NP_071531.1 | 2 | 1598 | ||
| XM_006232807.2 | XP_006232869.1 | 2 | 2130 | |||
| XM_008761206.1 | XP_008759428.1 | 3 | 1866 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ivl| Gene Symbol: Ivl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022195.1 | NP_071531.1 | 2 | 1598 | ||
| XM_006232807.2 | XP_006232869.1 | 2 | 2130 | |||
| XM_008761206.1 | XP_008759428.1 | 3 | 1866 | |||