siRNA Details
| Gene Symbol: Dusp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022199.1 | NP_071535.1 | 3 | 723 | ||
| GenBank mRNA | U23438.1 | - | 3 | 723 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp4| Gene Symbol: Dusp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022199.1 | NP_071535.1 | 3 | 723 | ||
| GenBank mRNA | U23438.1 | - | 3 | 723 | ||