siRNA Details
| Gene Symbol: Fgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022211.1 | NP_071547.1 | 2 | 415 | ||
| GenBank mRNA | D64085.1 | - | 2 | 415 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010203, BC071227, M30643, AK028894, AK028694.

Fgf5
Genomic Map