siRNA Details
| Gene Symbol: Insrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022212.2 | NP_071548.2 | 16 | 3277 | ||
| XM_006232749.2 | XP_006232811.2 | 15 | 3524 | |||
| XM_008761207.1 | XP_008759429.1 | 15 | 3524 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Insrr| Gene Symbol: Insrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022212.2 | NP_071548.2 | 16 | 3277 | ||
| XM_006232749.2 | XP_006232811.2 | 15 | 3524 | |||
| XM_008761207.1 | XP_008759429.1 | 15 | 3524 | |||