siRNA Details
| Gene Symbol: Cxcl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022214.1 | NP_071550.1 | 3 | 266 | ||
| XM_008770053.1 | XP_008768275.1 | 10 | 1663 | |||
| XM_008770054.1 | XP_008768276.1 | 3 | 683 | |||
| XM_008770055.1 | XP_008768277.1 | 10 | 1660 | |||
| GenBank mRNA | U90448.1 | - | 3 | 266 | ||

Cxcl6
Genomic Map