siRNA Details
| Gene Symbol: Cxcl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022214.1 | NP_071550.1 | 3 | 297 | ||
| GenBank mRNA | U90448.1 | - | 3 | 297 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcl6| Gene Symbol: Cxcl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022214.1 | NP_071550.1 | 3 | 297 | ||
| GenBank mRNA | U90448.1 | - | 3 | 297 | ||