siRNA Details
| Gene Symbol: Gulo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022220.2 | NP_071556.2 | 10 | 1118 | ||
| GenBank mRNA | BC089803.1 | - | 10 | 1118 | 7324344 | |
| FQ219140.1 | - | 10 | 1143 | |||
| FQ219632.1 | - | 10 | 1226 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gulo| Gene Symbol: Gulo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022220.2 | NP_071556.2 | 10 | 1118 | ||
| GenBank mRNA | BC089803.1 | - | 10 | 1118 | 7324344 | |
| FQ219140.1 | - | 10 | 1143 | |||
| FQ219632.1 | - | 10 | 1226 | |||