siRNA Details
| Gene Symbol: Per2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031678.1 | NP_113866.1 | 7, 8 | 922 | ||
| XM_006245476.2 | XP_006245538.1 | 7, 8 | 1157 | |||
| XM_006245477.2 | XP_006245539.1 | 7, 8 | 1073 | |||
| GenBank mRNA | AB016532.1 | - | 7, 8 | 922 | ||

Per2
Genomic Map