siRNA Details
| Gene Symbol: P2ry4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031680.1 | NP_113868.1 | 2 | 641 | ||
| XM_006257093.2 | XP_006257155.1 | 4 | 1281 | |||
| XM_006257094.2 | XP_006257156.1 | 3 | 850 | |||
| XM_006257095.2 | XP_006257157.1 | 3 | 1166 | |||
| XM_008773269.1 | XP_008771491.1 | 5 | 999 | |||
| XM_008773270.1 | XP_008771492.1 | 5 | 981 | |||
| GenBank mRNA | Y11433.1 | - | 2 | 641 | ||
More Information
Set Membership: |
|

P2ry4
Genomic Map