siRNA Details
| Gene Symbol: Pter | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022224.2 | NP_071560.2 | 3, 4 | 768 | ||
| XM_006254290.2 | XP_006254352.2 | 3, 4 | 4037 | |||
| XM_006254291.2 | XP_006254353.1 | 3, 4 | 855 | |||
| GenBank mRNA | BC078833.1 | - | 3, 4 | 768 | 7110694 | |

Pter
Genomic Map