siRNA Details
| Gene Symbol: Asic5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022227.1 | NP_071563.1 | 10 | 1454 | ||
| GenBank mRNA | Y19034.1 | - | 10 | 1454 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asic5| Gene Symbol: Asic5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022227.1 | NP_071563.1 | 10 | 1454 | ||
| GenBank mRNA | Y19034.1 | - | 10 | 1454 | ||