siRNA Details
| Gene Symbol: Ugt2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022228.1 | NP_071564.1 | 4 | 1013 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugt2a1| Gene Symbol: Ugt2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022228.1 | NP_071564.1 | 4 | 1013 | ||