siRNA Details
| Gene Symbol: Rapgef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233902.2 | XP_006233964.1 | 7 | 4052 | ||
| XM_008761683.1 | XP_008759905.1 | 6 | 969 | |||
| XM_008761684.1 | XP_008759906.1 | 6 | 969 | |||
| XM_008761685.1 | XP_008759907.1 | 5 | 855 | |||
| XM_008761686.1 | XP_008759908.1 | 5 | 855 | |||
| XM_008761687.1 | XP_008759909.1 | 6 | 969 | |||
| GenBank mRNA | AJ249986.1 | - | 4 | 508 | ||

Rapgef1
Genomic Map