siRNA Details
| Gene Symbol: Kcne3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022235.2 | NP_071571.1 | 2 | 364 | ||
| XM_006229785.2 | XP_006229847.1 | 3 | 2356 | |||
| XM_006229786.2 | XP_006229848.1 | 3 | 1211 | |||
| GenBank mRNA | AJ271742.1 | - | 3 | 499 | ||
| BC086406.1 | - | 2 | 364 | 7306835 | ||

Kcne3
Genomic Map