siRNA Details
| Gene Symbol: Abcb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022238.1 | NP_071574.1 | 4 | 1035 | ||
| GenBank mRNA | AB027520.1 | - | 4 | 1035 | ||
| AB116264.1 | - | 4 | 1035 | |||
| AB116265.1 | - | 4 | 1035 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcb9| Gene Symbol: Abcb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022238.1 | NP_071574.1 | 4 | 1035 | ||
| GenBank mRNA | AB027520.1 | - | 4 | 1035 | ||
| AB116264.1 | - | 4 | 1035 | |||
| AB116265.1 | - | 4 | 1035 | |||