siRNA Details
| Gene Symbol: Nmu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022239.2 | NP_071575.1 | 7, 8 | 561 | ||
| XM_006250872.2 | XP_006250934.1 | 6, 7 | 1351 | |||
| GenBank mRNA | M94555.1 | - | 7, 8 | 561 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nmu| Gene Symbol: Nmu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022239.2 | NP_071575.1 | 7, 8 | 561 | ||
| XM_006250872.2 | XP_006250934.1 | 6, 7 | 1351 | |||
| GenBank mRNA | M94555.1 | - | 7, 8 | 561 | ||