siRNA Details
| Gene Symbol: Arhgap17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270692.1 | NP_001257621.1 | 19, 20 | 2528 | ||
| NM_022244.2 | NP_071580.1 | 18, 19 | 2294 | |||
| GenBank mRNA | AB042827.1 | - | 18, 19 | 2293 | ||
| AB060557.1 | - | 19, 20 | 2527 | |||
| BC085736.1 | - | 19, 20 | 2529 | 7130218 | ||

Arhgap17
Genomic Map