siRNA Details
Gene Symbol: Smc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031683.1 | NP_113871.1 | 15 | 2404 | ||
GenBank mRNA | AJ005113.1 | - | 15 | 2404 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Smc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031683.1 | NP_113871.1 | 15 | 2404 | ||
GenBank mRNA | AJ005113.1 | - | 15 | 2404 |