siRNA Details
| Gene Symbol: Cyb5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022245.1 | NP_071581.1 | 4 | 374 | ||
| GenBank mRNA | AF007107.1 | - | 4 | 395 | ||
| AF007108.1 | - | 4 | 337 | |||
| BC086945.1 | - | 4 | 368 | 7322141 | ||
| D13205.1 | - | 4 | 374 | |||
| FQ209861.1 | - | 4 | 365 | |||
| FQ210790.1 | - | 4 | 362 | |||
| FQ211029.1 | - | 2 | 50 | |||
| FQ224354.1 | - | 4 | 365 | |||

Cyb5a
Genomic Map