siRNA Details
| Gene Symbol: Cyb5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022245.1 | NP_071581.1 | 2, 3 | 327 | ||
| GenBank mRNA | AF007107.1 | - | 2, 3 | 290 | ||
| AF007108.1 | - | 2, 3 | 290 | |||
| BC086945.1 | - | 2, 3 | 321 | 7322141 | ||
| D13205.1 | - | 2, 3 | 327 | |||
| FQ209861.1 | - | 2, 3 | 318 | |||
| FQ210790.1 | - | 2, 3 | 315 | |||
| FQ224354.1 | - | 2, 3 | 318 | |||

Cyb5a
Genomic Map