siRNA Details
| Gene Symbol: Pdcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022247.1 | NP_071583.1 | 4 | 678 | ||
| GenBank mRNA | BC097322.1 | - | 4 | 672 | 7453703 | |
| FQ211647.1 | - | 664 | ||||
| FQ223348.1 | - | 2076 | ||||
| L15354.1 | - | 4 | 678 | |||
| L15355.1 | - | 2 | 551 | |||
| Gene Symbol: LOC690155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761806.1 | XP_008760028.1 | 4 | 938 | ||

Pdcl
Genomic Map