siRNA Details
| Gene Symbol: Pdcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022247.1 | NP_071583.1 | 2, 3 | 291 | ||
| GenBank mRNA | BC097322.1 | - | 2, 3 | 285 | 7453703 | |
| FQ211647.1 | - | 277 | ||||
| L15354.1 | - | 2, 3 | 291 | |||
| Gene Symbol: LOC690155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761806.1 | XP_008760028.1 | 2, 3 | 551 | ||

Pdcl
Genomic Map