siRNA Details
| Gene Symbol: Khdrbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022249.1 | NP_071585.1 | 4, 5 | 583 | ||
| GenBank mRNA | AF152547.1 | - | 4, 5 | 583 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Khdrbs3| Gene Symbol: Khdrbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022249.1 | NP_071585.1 | 4, 5 | 583 | ||
| GenBank mRNA | AF152547.1 | - | 4, 5 | 583 | ||