siRNA Details
| Gene Symbol: Pag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022253.1 | NP_071589.1 | 985 | |||
| GenBank mRNA | AB038038.1 | - | 985 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pag1| Gene Symbol: Pag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022253.1 | NP_071589.1 | 985 | |||
| GenBank mRNA | AB038038.1 | - | 985 | |||