siRNA Details
| Gene Symbol: Masp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022257.1 | NP_071593.1 | 9 | 1272 | ||
| XM_006248526.2 | XP_006248588.1 | 9 | 1516 | |||
| GenBank mRNA | AJ277423.1 | - | 8 | 1168 | ||
| AJ457084.1 | - | 9 | 1272 | |||
| AJ487622.1 | - | 8 | 1068 | |||
| AJ487624.1 | - | 3 | 259 | |||

Masp1
Genomic Map