siRNA Details
| Gene Symbol: Pdcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022265.2 | NP_071601.2 | 6 | 839 | ||
| XM_006231613.2 | XP_006231675.1 | 7 | 1120 | |||
| XM_006231614.2 | XP_006231676.1 | 7 | 1172 | |||
| XM_006231615.2 | XP_006231677.1 | 7 | 1018 | |||
| XM_006231616.2 | XP_006231678.1 | 7 | 995 | |||
| GenBank mRNA | AF239739.2 | - | 6 | 839 | ||
| BC167751.1 | - | 7 | 1027 | 9027822 | ||

Pdcd4
Genomic Map