siRNA Details
| Gene Symbol: Ctgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022266.2 | NP_071602.1 | 5 | 983 | ||
| GenBank mRNA | AB023068.1 | - | 5 | 971 | ||
| AF079531.1 | - | 2 | 117 | |||
| AF120275.1 | - | 5 | 983 | |||
| AY596447.1 | - | 1 | 1544 | |||
| BC072503.1 | - | 4 | 1352 | 7098774 | ||

Ctgf
Genomic Map