siRNA Details
| Gene Symbol: Aldh9a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022273.2 | NP_071609.2 | 7 | 972 | ||
| GenBank mRNA | AF170918.1 | - | 7 | 959 | ||
| BC074019.1 | - | 7 | 972 | 7104823 | ||
| Gene Symbol: LOC100910278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250207.2 | XP_006250269.1 | 1 | 135 | ||

Aldh9a1
Genomic Map