siRNA Details
| Gene Symbol: Gcnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022276.1 | NP_071612.1 | 1 | 1451 | ||
| XM_006231147.2 | XP_006231209.1 | 3 | 1060 | |||
| XM_006231150.2 | XP_006231212.1 | 3 | 1152 | |||
| GenBank mRNA | S79797.1 | - | 1 | 1451 | ||

Gcnt1
Genomic Map