siRNA Details
| Gene Symbol: Glrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022278.1 | NP_071614.1 | 2 | 287 | ||
| GenBank mRNA | AF167981.3 | - | 2 | 287 | ||
| AF319950.1 | - | 2 | 256 | |||
| BC061555.1 | - | 2 | 316 | 6889009 | ||
| FQ224532.1 | - | 2 | 323 | |||

Glrx
Genomic Map