siRNA Details
Gene Symbol: Lrat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022280.2 | NP_071616.1 | 1 | 637 | ||
GenBank mRNA | AF255060.2 | - | 1 | 637 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022280.2 | NP_071616.1 | 1 | 637 | ||
GenBank mRNA | AF255060.2 | - | 1 | 637 |