siRNA Details
| Gene Symbol: Guca2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022284.2 | NP_071620.1 | 3 | 454 | ||
| GenBank mRNA | U41322.1 | - | 3 | 454 | ||
| U73898.1 | - | 3 | 471 | |||
| U75186.1 | - | 3 | 581 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Guca2b| Gene Symbol: Guca2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022284.2 | NP_071620.1 | 3 | 454 | ||
| GenBank mRNA | U41322.1 | - | 3 | 454 | ||
| U73898.1 | - | 3 | 471 | |||
| U75186.1 | - | 3 | 581 | |||