siRNA Details
| Gene Symbol: Slc26a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022287.2 | NP_071623.2 | 3 | 1027 | ||
| GenBank mRNA | BC085735.1 | - | 3 | 1027 | 7108820 | |
| L23413.1 | - | 3 | 1349 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc26a1| Gene Symbol: Slc26a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022287.2 | NP_071623.2 | 3 | 1027 | ||
| GenBank mRNA | BC085735.1 | - | 3 | 1027 | 7108820 | |
| L23413.1 | - | 3 | 1349 | |||