siRNA Details
| Gene Symbol: Csnk1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022288.1 | NP_071624.1 | 938 | |||
| XM_008766332.1 | XP_008764554.1 | 5 | 661 | |||
| XM_008766333.1 | XP_008764555.1 | 5 | 661 | |||
| XM_008766334.1 | XP_008764556.1 | 5 | 661 | |||
| XM_008766335.1 | XP_008764557.1 | 5 | 661 | |||
| GenBank mRNA | BC078831.1 | - | 940 | 7134461 | ||
| U22296.1 | - | 938 | ||||

Csnk1g1
Genomic Map