siRNA Details
| Gene Symbol: Snx16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022289.3 | NP_071625.2 | 4, 5 | 907 | ||
| XM_006232135.2 | XP_006232197.1 | 4, 5 | 920 | |||
| GenBank mRNA | AF305780.1 | - | 4, 5 | 893 | ||
| BC061554.1 | - | 4, 5 | 911 | 6890629 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: AK183064, AK015872, AK032055, BC019424, AK166422, and others.

Snx16
Genomic Map