siRNA Details
| Gene Symbol: Kcnk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022292.1 | NP_071628.1 | 1326 | |||
| GenBank mRNA | AF287300.1 | - | 1326 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnk12| Gene Symbol: Kcnk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022292.1 | NP_071628.1 | 1326 | |||
| GenBank mRNA | AF287300.1 | - | 1326 | |||