siRNA Details
| Gene Symbol: Scn7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031686.1 | NP_113874.1 | 25 | 4902 | ||
| XM_006234296.2 | XP_006234358.1 | 25 | 5113 | |||
| XM_006234298.2 | XP_006234360.1 | 24 | 5005 | |||
| GenBank mRNA | M96578.1 | - | 2 | 1091 | ||
| Y09164.1 | - | 25 | 4902 | |||

Scn7a
Genomic Map