siRNA Details
| Gene Symbol: Ddah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022297.2 | NP_071633.1 | 1, 2 | 413 | ||
| GenBank mRNA | FQ220049.1 | - | 1, 2 | 433 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddah1| Gene Symbol: Ddah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022297.2 | NP_071633.1 | 1, 2 | 413 | ||
| GenBank mRNA | FQ220049.1 | - | 1, 2 | 433 | ||