siRNA Details
| Gene Symbol: Card9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022303.1 | NP_071639.1 | 11 | 1517 | ||
| GenBank mRNA | AF311288.1 | - | 11 | 1517 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Card9| Gene Symbol: Card9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022303.1 | NP_071639.1 | 11 | 1517 | ||
| GenBank mRNA | AF311288.1 | - | 11 | 1517 | ||